Learn More
Heathrow Scientific™ Cardboard Cryogenic Tube Storage Box and Lid
GREENER_CHOICE
Catalog No. 17100439
Change view
Click to view available options
Color:
Assorted
Beige
Blue
Red
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-439 Beige
17-100-443 White
17-100-456 Assorted
17-100-447 Blue
17-100-423 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-100-439 Supplier Heathrow Scientific™ Supplier No. HS2860B
Add to Cart
Add to Cart
Cardboard Cryogenic Tube Storage Box and Lid for storage of small samples and tubes.
  • Made from cardboard
  • Colors allow for easy identification and sorting purposes
Specifications
Color Beige
Material Cardboard
Shape Square
Type Cryo Box
Holds 1.5/2.0 mL Tubes
Withstand Temperature (Metric) -196°C
Dimensions (L x W x H) 5.2 x 5.2 x 3 in. (13.3 x 13.3 x 7.5 cm)