Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardiac Leiomodin Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB293733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293733 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293733 Supplier Novus Biologicals Supplier No. NBP326068AF532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cardiac Leiomodin Polyclonal antibody specifically detects Cardiac Leiomodin in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Cardiac Leiomodin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C-LMOD, Leiomodin 2 (Cardiac), leiomodin-2, LMOD2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to Human Cardiac Leiomodin
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442721
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.