Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardiac Leiomodin Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326068AF647
Description
Cardiac Leiomodin Polyclonal antibody specifically detects Cardiac Leiomodin in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Cardiac Leiomodin | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
C-LMOD, Leiomodin 2 (Cardiac), leiomodin-2, LMOD2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to Human Cardiac Leiomodin | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
442721 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction