Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH7B Recombinant Rabbit Monoclonal Antibody (JE54-44)
GREENER_CHOICE

Catalog No. PIMA536186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536186 Supplier Invitrogen™ Supplier No. MA536186
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MYH7B Recombinant Monoclonal Antibody for Western Blot

Myosin cardiac muscle beta chain or MYH7B is believed to be involved in cardiac slow-twitch myosin contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH7B
Applications Western Blot
Classification Recombinant Monoclonal
Clone JE54-44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MYH7B
Gene Accession No. A2AQP0, A7E2Y1
Gene Alias A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-cardiac myosin heavy chain; alpha-MHC; Antigen MLAA-21; ASD3; beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin heavy chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain alpha isoform; cardiac myosin heavy chain beta isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMD1S; CMH1; CMH14; KIAA1512; LOC100736765; MHC14; MPD1; Myh14; MYH6; MYH7; Myh7b; MYH-beta/slow; MYHC; Myhca; Myhc-a; MyHC-alpha; Myhcb; Myhc-b; MyHC-beta; MyHC-I; myhc-slow; myopathy, distal 1; myosin 7; Myosin cardiac muscle beta chain; Myosin heavy chain 6; myosin heavy chain 7; myosin heavy chain 7B; myosin heavy chain 7B, cardiac muscle beta isoform; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, polypeptide 7; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy chain 7B, cardiac muscle, beta; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; myosin, heavy polypeptide 7B, cardiac muscle, beta; myosin-6; Myosin-7; myosin-7B; rhabdomyosarcoma antigen MU-RMS-40.7A; Slow A MYH14; slow myosin heavy chain-beta; SPMD; SPMM; SSS3; type slow/beta myosin heavy chain; U937-associated antigen; Unknown (protein for IMAGE:7985957)
Gene Symbols MYH7B
Host Species Rabbit
Immunogen Recombinant protein within Human MYH7B aa 1,710-1,983.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311570, 57644, 668940
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.