Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin C Monoclonal Antibody (7B9)
GREENER_CHOICE

Catalog No. PIMA122698
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA122698 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA122698 Supplier Invitrogen™ Supplier No. MA122698
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-22698 detects Cardiac Troponin C in Human samples.

Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin C
Applications Affinity Purification, ELISA, Western Blot
Classification Monoclonal
Clone 7B9
Concentration 6.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Tnnc1
Gene Accession No. P63316
Gene Alias AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; Tncc; Tni; TNN I3; TNNC; TNNC 1; Tnnc1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI
Gene Symbols Tnnc1
Host Species Mouse
Immunogen Human cardiac troponin complex.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7134
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.