Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116462 Supplier Invitrogen™ Supplier No. PA5116462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls: Human heart tissue lysates, MG-63, SKOV-3, SW620, rat skeletal muscle tissue, rat heart tissue, mouse skeletal muscle tissue, mouse heart tissue.

Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin C
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Tnnc1
Gene Accession No. P19123, P63316
Gene Alias AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; Tncc; Tni; TNN I3; TNNC; TNNC 1; Tnnc1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI
Gene Symbols Tnnc1
Host Species Rabbit
Immunogen Recombinant protein within Human TNNC1 aa 1-161.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21924, 290561, 7134
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.