Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin T Recombinant Rabbit Monoclonal Antibody (17H8L13)

Catalog No. 701620
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-162-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-162-0 Supplier Invitrogen™ Supplier No. 701620
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin T
Applications Immunocytochemistry
Classification Recombinant Monoclonal
Clone 17H8L13
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene TNNT2
Gene Accession No. P45379
Gene Alias alternatively spliced; cardiac muscle troponin T; cardiac TnT; cardiac troponin 2; cardiac troponin 2 adult #1 isoform; cardiac troponin 2 adult #2 isoform; cardiac troponin 2 embryonic #1 isoform; cardiac troponin 2 embryonic #2 isoform; cardiac troponin 2, exclusion exon 6 #1 isoform; cardiac troponin 2, exclusion exon 6 #2 isoform; cardiac troponin T; cardiac troponin T form I; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T isoform I; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, hypertrophic 2; CMD1D; CMH2; CMPD2; cTnT; Ctt; CTTG; LVNC6; MGC3889; RATCTTG; RCM3; TNNT2; Tnnt3; Tnt; TnTc; troponin T; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin T splicing mutant isoform 2; troponin T type 2 (cardiac); troponin T, cardiac muscle; Troponin T, cardiac muscle isoforms; Troponin T2; troponin T2, cardiac; troponin T2, cardiac type; truncated cardiac troponin T
Gene Symbols TNNT2
Host Species Rabbit
Immunogen human TNNT2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.