Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardinal Health™ General-Purpose Coveralls With Tyvek™
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Description
- Feature full-length front zipper and elastic wrists and ankles
- Basic style offers set-in sleeves and traditional collar
- Premier style offers raglan sleeves, bound collar, and a more generous cut
- Both styles are white
Order Info
To ensure cleanliness, coveralls are packed in double poly-lined bags.

Specifications
Specifications
Product Type | Cleanroom Coveralls |
Size | 2X-Large |
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
For Use With (Application) | Controlled Environments |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction