Learn More
Cardinal Health™ Low-Temperature Sterilization Pouchess
Catalog No. p-3752536 Shop All Cardinal Health Products
Click to view available options
Dimensions (L x W):
15.2 x 25.4cm (6 x 10 in.)
20.3 x 30.5cm (8 x 12 in.)
25.4 x 38.1cm (10 x 15 in.)
7.6 x 17.9cm (3 x 7 in.)
Closure Type:
Self-seal
This item is not returnable. View return policy
This item is not returnable. View return policy
Protect the sterile integrity of tools and equipment
For use with low-temperature sterilization and applications requiring EO.
  • Breathable Tyvek™ material facilitates faster aeration and easier sterilization
  • Resistant to puncture and tearing
Specifications
Closure Type Self-seal
For Use With (Application) Excellent choice for devices requiring low temperature sterilization, produced without an indicator for all types of low temperature sterilization, including EO and plasma
Material Tyvek
Dimensions (L x W) 25.4 x 38.1cm (10 x 15 in.)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.