Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTS Diagnostics CardioCheck™ Plus Analyzer
Click to view available options
Detects:
HDL, LDL, Triglycerides, Total Cholesterol
Description
- Measures total cholesterol, HDL cholesterol, triglycerides and glucose
- Calculates LDL, TC/HDL ratio, LDL/HDL ratio, and non-HDL cholesterol
- Large display with two button operation
- Improved bezel design
- MEMo Chip™ port with guides for ease of insertion
- With USB connection port, electrochemical glucose test strip port and reflectance test strip slot
- Compatible with all existing PTS Panels™ test strips
- Wi-Fi communication compatible
- Smart Bundle Pack includes eGLU test strips (21124412)
- Accessories available; see cat. nos. 21124411 - 21124412
Specifications
Specifications
| Type | CardioChek Plus Analyzer |
| Sample Type | Whole Blood |
| For Use With (Application) | Measuring cholesterol and glucose with whole blood samples |
| Analysis Time | 10 sec. eGLU; 2 min. Lipids |
| Detects | HDL, LDL, Triglycerides, Total Cholesterol |
| Sample Size | 15 μL |
| Includes | 1 CardioChek Plus Analyzer, User Guide / Quick Reference Guide, Warranty Card,4 AA Batteries |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction