Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer without WiFi
SDPSCI_ED

Catalog No. p-7184562 Shop All 3B Scientific Products
Click to view available options
Color:
Dark Skin
Light Skin

3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer without WiFi uses a SimScope stethoscope to simulate pediatric heart, lung, and bowel sounds

  • The PAT Basic™ trainer engages the technology of the SimScope stethoscope to simulate pediatric heart, lung, and bowel sounds utilizing a large sounds library
  • Using the provided SimScope stethoscope, users are able to access a large variety of pathological conditions in order to customize scenarios
  • By placing the SimScope over the anatomically correct locations of PAT Basic™, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills
  • PAT Basic™ consists of a Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual

Specifications

Product Type Manikin
For Use With (Application) Teaching or Learning Pediatric Heart, Lung, and Bowel Sounds
Color Light Skin
Includes Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual
Weight (English) 9.91 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.