Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Cardionics PAT™ Pediatric Auscultation Trainer
SDPSCI_ED

Catalog No. S27942 Shop All 3B Scientific Products
Click to view available options
Color:
Dark Skin
Light Skin

3B Scientific™ Cardionics PAT™ Pediatric Auscultation Trainer is the complete auscultation manikin for pediatric-specific conditions

  • PAT is the complete auscultation manikin for pediatric-specific conditions
  • Since pediatric care requires careful consideration of growth and developmental issues, PAT is designed to prepare students to readily recognize the signs and sounds associated with pediatric-specific disorders
  • With one of the largest libraries of pediatric sounds and intuitive instructional tools, PAT gives students everything they need to actively engage in their auscultation education
  • Pediatric size auscultation trainer with listening points at the correct anatomical locations
  • Heart sounds at different rates for comparison, (e.g. Example: Atrial Septal Defect at 75 bm and 90 bm)
  • PAT's computer software interface is easily projected into any smart classroom
  • The software includes phonocardiograms, correct anatomical locations, and written lessons for each sound
  • With programmable and password-protected lectures, many institutions find value in having numerous instructors utilize PAT

Specifications

Product Type Manikin
For Use With (Application) Teaching or Learning Pediatric-Specific Diseases
Color Light Skin
Weight (English) 22.93 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.