Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Cardionics PAT™ Pediatric Auscultation Trainer

Shop All 3B Scientific Products


Click to view available options
Color:
Dark Skin
Light Skin
Description
- PAT is the complete auscultation manikin for pediatric-specific conditions
- Since pediatric care requires careful consideration of growth and developmental issues, PAT is designed to prepare students to readily recognize the signs and sounds associated with pediatric-specific disorders
- With one of the largest libraries of pediatric sounds and intuitive instructional tools, PAT gives students everything they need to actively engage in their auscultation education
- Pediatric size auscultation trainer with listening points at the correct anatomical locations
- Heart sounds at different rates for comparison, (e.g. Example: Atrial Septal Defect at 75 bm and 90 bm)
- PAT's computer software interface is easily projected into any smart classroom
- The software includes phonocardiograms, correct anatomical locations, and written lessons for each sound
- With programmable and password-protected lectures, many institutions find value in having numerous instructors utilize PAT
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Teaching or Learning Pediatric-Specific Diseases |
| Color | Light Skin |
| Weight (English) | 22.93 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction