Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Cardionics SAM II™ Student Auscultation Manikin

Shop All 3B Scientific Products


Click to view available options
Color:
Dark Skin
Light Skin
Description
Product Features:
- Auscultation is an essential clinical skill needed to assess and monitor patients' conditions
- The SAM II™ trainer can be used with any stethoscope
- All sounds and videos are recorded from live patients for students to experience a life-like simulation
- Users can also create and save their own case videos
- When connected to the included laptop with pre-installed software, a variety of sounds, videos and lessons recorded can be accessed
- When connected to speakers (Sold Separately) SAM II Student Auscultation Manikin can be easily moved into a classroom or an auditorium for group instruction
- The programmable and password protected lectures allow teaching institutions to have numerous instructors utilize the SAM II trainer
Delivery content:
- Portable SAM II Male Auscultation Torso
- New DELL laptop computer pre-installed with auscultation teaching software, programmable and password protected lectures
- Digital copy of SAM`s Lesson Guide: located on the laptop in digital form, can be printed and used to support auscultation teaching anytime
- One SAM t-shirt
- User's manual
How is SAM II Student Auscultation Manikin different from other auscultation products available on the market?
- It is lightweight & portable
- It has the largest library of sounds (24 sounds) & videos (36 videos altogether)
- It has the complete Lesson Guide
- It can be used with any stethoscope
- It has a palpable carotid pulse
- Users can setup password protected lectures for student assessment
- It is available in light and dark skin version
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Teaching and Learning Auscultation |
| Color | Light Skin |
| Includes | SAM II Male Auscultation Torso, Dell Laptop with Pre-Installed Auscultation Teaching Software, SAM's Lesson Guide, SAM T-Shirt, User's Manual |
| Weight (English) | 30.20 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction