Description
Includes
Cytonin™, 5mL; Proteinase K, 30uL; TACS Blue Label, 1.5mL; TACS-Nuclease, 15uL; Red Counterstain C, 50mL; Strep-HRP, 30uL; Also includes TdT enzyme, buffers and dNTPs, 50X Mn+2; Other reagents
Apoptosis is characterized by a number of intracellular phenomena such as membrane blebbing, chromatin condensation and nuclear DNA fragmentation. Detection of nuclear DNA fragmentation is a widely accepted method to assay for apoptosis and can be perform
Complete kits contain TACS BlueLabel™, Nuclear Fast Red counterstain and reagents necessary for generating positive controls
Kits are optimized for detection of apoptosis in cardiac tissues
Study fixed cells, embedded tissues and froze
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 30 Tests |
| Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Product Line | R&D Systems |