Learn More
Invitrogen™ Cardiotin Monoclonal Antibody (R2G)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ OMA106030
Description
OMA1-06030 detects cardiotin from mouse, rat, hamster, canine, monkey, pig, cat, rabbit, frog, goat, xenopus,feline and human cardiomyocytes. OMA1-06030 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~300 kDa protein representing the cardiotin protein complex, as well as ~100 kDa and ~60 kDa bands representing the cardiotin subunits from rat heart extract. The OMA1-06030 immunogen is total protein extract from chicken gizzard.
Cardiotin is a recently discovered, high molecular weight protein complex (~300 kDa) located in the longitudinal SR of cardiomyocytes. The cardiotin complex exists as 60 kDa and 100 kDa subunits. Both subunits contain the same amino-terminal 14 amino-acid sequence, and show high homology to human skeletal muscle alpha-actin. The tetrameric configuration of the cardiotin protein complex suggests that it exists as a transmembrane complex with the N-terminus at the cytoplasmic side of the membrane, able to interact with actin. Immunofluorescence studies colocalize cardiotin with SERCA 2a ATPase in the longitudinal sarcoplasmic reticulum (SR), although cardiotin is not expressed in junctional SR with SERCA 2.
Specifications
Cardiotin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Total protein extract of chicken gizzard. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
R2G | |
Unconjugated | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Human, Mouse, Rat, Canine, Rabbit, Monkey, Hamster, Feline, Chicken, Frog, Zebrafish, Goat | |
Antibody | |
IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.