Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Cards, Makerspace 6-8, with Ring, Set of 72

Click to view available options
Class Size:
1 Class
Description
- These cards provide structure by giving students ideas about what to build and how to challenge themselves to improve their designs.
- Each card provides design ideas with real-world pictures, tips to get the students thinking, and suggested modifications to further challenge the students with their design.
Specifications
Specifications
Grade Level | Intermediate - Middle School |
Grade | 6 to 8 |
Class Size | 1 Class |
Product Type | Makerspace Cards |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction