Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Cards, with Ring, Set of 36

Click to view available options
Grade:
3 to 5
K to 2
Description
- These cards provide structure by giving students ideas about what to build and how to challenge themselves to improve their designs.
- Each card includes design ideas with real-world pictures, tips to get the students thinking, and suggested modifications to further challenge the students with their design.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | K to 2 |
| Class Size | 1 Class |
| Product Type | STEAM Learning |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction