Learn More
Invitrogen™ CARF Polyclonal Antibody

Description
The antibody detects endogenous levels of total CARF / ALS2CR8 protein.
Specifications
Specifications
| Antigen | CARF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CARF |
| Gene Accession No. | Q8N187 |
| Gene Alias | Als2cr8; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein homolog; calcium response factor; calcium responsive transcription factor; calcium-response factor; calcium-response factor CaRF; calcium-responsive transcription factor; CaRF; ECBRC-FC1; ECBRC-FC2; NYD-SP24; testis development protein NYD-SP24 |
| Gene Symbols | CARF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.