Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARHSP1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300026PE
Description
CARHSP1 Polyclonal antibody specifically detects CARHSP1 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
CARHSP1 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
calcium regulated heat stable protein 1 (24kD), calcium regulated heat stable protein 1, 24kDa, calcium-regulated heat stable protein 1, calcium-regulated heat-stable protein (24kD), Calcium-regulated heat-stable protein of 24 kDa, CRHSP-24MGC111446, CSDC1 | |
Produced in rabbits immunized with purified, recombinant Human CARHSP1 (Uniprot#: Q9Y2V2; Met1-Ser147) | |
0.1 mL | |
Signal Transduction | |
23589 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction