Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cariporide
GREENER_CHOICE

Catalog No. 535810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-581-0 10 mg
53-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-581-0 Supplier Tocris Bioscience™ Supplier No. 5358/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective NHE1 inhibitor; cardioprotective and antitumor

Cariporide is a selective Na+/H+ exchanger isoform 1 (NHE1) inhibitor (IC50 values are 0.05, 3 and 1000 μM for NHE1, NHE3 and NHE2 respectively). Attenuates ischemia-induced cardiomyocyte apoptosis in vitro. Reduces cardiac arrhythmia in vivo. Also promotes apoptosis in cancer cells overexpressing NHE1. Orally active.

Specifications

Quantity 10 mg
Product Type Cariporide
Purity >99%
Molecular Weight (g/mol) 283.35
Inhibitors Cariporide

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.