Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARKL Antibody (OTI3D7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271771G
Description
CARKL Monoclonal specifically detects CARKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CARKL | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SHPK | |
Full length recombinant protein of human CARKL (NP_037408) produced in HEK293T cells. | |
0.1 mL | |
23729.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3D7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
carbohydrate kinase-like, Carbohydrate kinase-like protein, CARKLEC 2.7.1.14, sedoheptulokinase, SHKFLJ32478 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction