Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARKL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564191 Supplier Invitrogen™ Supplier No. PA564191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILQALHECLA ALPRPQLRSV VGIGVSGQMH GVVFWKTGQG CEWTEGGITP VFEPRAVSHL VTWQDGRCSS EFLASLPQPK SHLSVATGFG CATIFWLLKY R Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARKL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHPK
Gene Accession No. Q9UHJ6
Gene Alias 4930431K22Rik; AI194947; AW260459; Carbohydrate kinase-like protein; CARKL; Sedoheptulokinase; SHK; Shpk
Gene Symbols SHPK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CARKL. Recombinant protein control fragment (Product #RP-103945).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.