Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carm1 Antibody (3H2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237645
Description
Carm1 Monoclonal specifically detects Carm1 in Human, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Carm1 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
CARM1 | |
Purified recombinant fragment of human CARM1 expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human, Rat, Monkey, Primate | |
Ascites |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3H2 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
Mouse | |
65 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Transcription Factors and Regulators, Tumor Suppressors | |
10498 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction