Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carm1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200342H
Description
Carm1 Polyclonal antibody specifically detects Carm1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Carm1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
A synthetic peptide that maps to a region between residues 550 and the C-terminus (residue 585) of human Coactivator-Associated Arginine Methyltransferase 1 using the numbering given in Swiss-Prot entry Q86X55 (GeneID 10498). | |
0.1 mL | |
Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
10498 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction