Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carm1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321326100UL
Description
Carm1 Polyclonal antibody specifically detects Carm1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Carm1 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: NMWNTGSTYNLSSGMAVAGMPTAYDLSSVIASGSSVGHNNLIPLGSSGAQGSGGGSTSAHYAVNSQFTMGGPAISMASPMSIPTNTMHYGS | |
100 μg | |
Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
10498 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction