Learn More
CARM1 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | CARM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 30% glycerol and 0.035% sodium azide; pH |
| Gene | Carm1 |
| Gene Accession No. | Q4AE70, Q86X55, Q9WVG6 |
| Gene Alias | Carm1; coactivator associated arginine methyltransferase 1; Coactivator-associated arginine methyltransferase 1; coactivator-associated arginine methyltransferase 1 variant 5; Histone-arginine methyltransferase CARM1; PRMT4; protein arginine methyltransferase; Protein arginine N-methyltransferase 4 |
| Gene Symbols | Carm1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.