Learn More
CARMIL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304556AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Binds CAPZA2 with high affinity and significantly decreases CAPZA2 affinity for actin barbed ends. Increases the rate of elongation from seeds in the presence of CAPZA2, however, seems unable to nucleate filaments. Rapidly uncaps barbed ends capped by CAPZA2 and enhances barbed-end actin polymerization. May control actin dynamics in lamellipodia. Required for cell migration.Specifications
CARMIL | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5VZK9 | |
CARMIL1 | |
Between 1321 and 1371 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CARMIL1 | |
Leucine-rich repeat-containing protein 16A, CARMIL homolog, LRRC16A, CARMIL, CARMIL1a, LRRC16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55604 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.