Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmine, MP Biomedicals™

Catalog No. ICN15263405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15263405 5 g
ICN15263401 1 g
ICN15263425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15263405 Supplier MP Biomedicals, Inc Supplier No. 0215263405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1390-65-4
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-UHFFFAOYNA-N
Synonym Carminic acid, Cochineal, Natural Red 4, Nacarat
PubChem CID 25113282
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O

Specifications

Formula Weight 1150.87
Color Pink to Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Carmine
EINECSNumber 215-724-4
Recommended Storage Store at Room Temperature (15-30°C).
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.