Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnauba wax

Catalog No. AA31117A1
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 8015-86-9

Carnauba wax is used as a glazing agent, bulking agent, acidity regulator and tablet-coating agent. It is also used in automobile waxes, shoe polishes, dental floss and food products such as sweets, instrument polishes, floor, furniture waxes, polishes and paper coatings. It serves as the main ingredient in surfboard wax. Further, it is also used as a mold release agent to produce fiber-reinforced plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8015-86-9
MDL Number MFCD00130724
Synonym Wax

Specifications

Melting Point 82°C to 86°C
Density 0.990g/mL to 0.999g/mL
Odor Odorless
Refractive Index 1.45
Quantity 1 kg
Merck Index 14,1846
Solubility Information Soluble in diethyl ether,alkali,chloroform. Slightly soluble in boiling ethanol. Insoluble in water.
Physical Form Flakes
Chemical Name or Material Carnauba wax
EINECSNumber 232-399-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.