Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARNMT1 Antibody (OTI4B7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202664
Description
CARNMT1 Monoclonal specifically detects CARNMT1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
C9orf41 | |
Monoclonal | |
0.87 mg/mL | |
Western Blot 1:1000-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
CARNMT1 | |
Full length human recombinant protein of human C9orf41 (NP_689663) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
138199 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4B7 | |
Unconjugated | |
chromosome 9 open reading frame 41, FLJ25795, hypothetical protein LOC138199 | |
Mouse | |
47 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction