Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARNMT1 Antibody (OTI4B7), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272398PEATT594
Description
CARNMT1 Monoclonal specifically detects CARNMT1 in Human samples. It is validated for Flow Cytometry.Specifications
CARNMT1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
CARNMT1 | |
Full length recombinant protein of human C9orf41 (NP_689663) produced in HEK293T cells. | |
0.1 mL | |
138199.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI4B7 | |
Flow Cytometry | |
C9orf41, chromosome 9 open reading frame 41, FLJ25795, hypothetical protein LOC138199 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction