Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carnosic Acid (Synthetic) 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
20 mg
100 mg
Specifications
Specifications
| Melting Point | 190°C |
| Color | Yellow |
| Quantity | 100 mg |
| Formula Weight | 332.44 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Carnosic Acid (Synthetic) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction