Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carnosine Dipeptidase 1/CNDP1 Antibody (CL0339), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP23097225UL
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot.Specifications
Carnosine Dipeptidase 1/CNDP1 | |
Monoclonal | |
Unconjugated | |
Q96KN2 | |
CNDP1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PALLEKVFQYIDLHQDEFVQTLKEWVAIESDSVQPVPRFRQELFRMMAVAADTLQRLGARVASVDMGPQQLPDGQSLPIPPVILAELGSDPTKGTVCFYGHL | |
25 μL | |
Signal Transduction | |
84735 | |
Human | |
Purified |
Western Blot | |
CL0339 | |
Western Blot 1 ug/ml | |
beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction