Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carnosine Dipeptidase 1/CNDP1 Antibody (OTI2F8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147702
Description
Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carnosine Dipeptidase 1/CNDP1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
CNDP1 | |
Full length human recombinant protein of human CNDP1 (NP_116038) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens carnosine dipeptidase 1 (metallopeptidase M20 family) (CNDP1). | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI2F8 | |
Unconjugated | |
beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase | |
Mouse | |
56.7 kDa | |
0.1 mL | |
Signal Transduction | |
84735 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction