Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Carnoy's Solution
SCI_ED

Catalog No. s25237 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is for Educational use only and is not FDA approved for medical use.
This item is not returnable. View return policy
This item is for Educational use only and is not FDA approved for medical use.

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Carnoy's Solution
CAS 64-19-7, 67-66-3, 64-17-5
Density 1g/mL
Packaging 10% w/v
Quantity 100 mL
Assay Percent Range 10% w/v
Synonym Carnoy-Lebrun Solution (Fixative)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.