Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Butterflies in the Classroom Kit
SDPSCI_ED

Numéro de catalogue. p-4655582
missing translation for 'orderingAttributeHoverText'
Includes:
Live Caterpillars
Prepaid Coupon for Caterpillars
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Comprend

  • Painted Lady Butterfly Culture (33 larvae with food)
  • Painted Lady Butterfly Culture Medium (food)
  • Carolina Butterfly Sanctuary
  • 33 1 oz. cups with lids
  • Five wooden craft sticks
  • Five sorting brushes
  • 10 Kimwipes™
  • Foam flower
  • Cotton wick
  • Teacher's Manual and Reproducible Student Guide

Exige

Spray bottle; small bottle or vial; sugar; water; food coloring (optional); host plants (mallow, thistle, daisy, etc. optional)

These kits make keeping live butterflies simple, fun and affordable.

This kit allows students to work hands-on with living butterflies. From a culture cup that contains 33 larvae and food, each student (or pair of students) transfers a larva and food to its own culture vessel, creating an individual chamber for observation. The provided habitat is simple to set up, easy to clean, great for viewing and collapses for convenient storage.

Note: Prepaid coupons allow you to order your organisms in advance. We' ll send a coupon(s) that you later redeem by phone, fax, online Web form or mail. To ensure prompt arrival of material, coupons must be redeemed at least two weeks prior to requested delivery date.

Spécifications

For Use With (Application) Allow students to work hands on with living butterflies.
Quantity Ea.
Includes Prepaid Coupon for Caterpillars
Sufficient For Class of 30 students
Product Type Butterflies in the Classroom Kit (with Live Caterpillars)
missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'