Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Earthworms



Click to view available options
Quantity:
100/Container
12/Container
25/Container
Description
- Lumbricus terrestris
- Large, mature specimens are suitable for laboratory study and dissection
- Medium-sized specimens are not suitable for dissection, but are good for making slides and for use as food for snakes and large salamanders
- With instructions
Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.
Specifications
Specifications
For Use With (Application) | Laboratory study and dissection |
Storage Requirements | 15°C |
Quantity | 25/Container |
Size | Large |
Scientific Name | Lumbricus terrestris |
Includes | Instructions |
Product Type | Earthworm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction