Learn More
Carolina™ Carolina™ Formalin Grasshoppers
SDPSCI_ED
Catalog No. s72459 Shop All Carolina Biological Supply Products
Click to view available options
Injection Type:
Plain
Single
This item is not returnable. View return policy
This item is not returnable. View return policy
Carolina™ Formalin Grasshoppers are an excellent introductory specimen for dissecting and studying insect anatomy.
  • Louisiana Lubber Grasshoppers (Romalea microptera or Brachystola)
  • Perfect introductory dissection specimen
  • Large size is perfect for studying specialized external and internal anatomy
  • Preserved in formalin
  • Shipped 10 per jar
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid which minimizes student and educator exposure to formaldehyde
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 10/Jar
Packaging Jar
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.