Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Green Algae Mixture

Shop All Carolina Biological Supply Products


Click to view available options
Quantity:
Ea.
Description
Includes
- Chlorella
- Scenedesmus
- Selenastrum
- Ulothrix
- Volvox
- Materials for a class of 30 students
- Five representatives of green algae
Specifications
Specifications
| Storage Requirements | 22°C |
| Quantity | Ea. |
| Includes | Five representatives of Green Algae Chlorella, Scenedesmus, Selenastrum, Ulothrix, Volvox |
| Sufficient For | Class of 30 students |
| Product Type | Green Algae Mixture |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction