Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Anolis carolinensis
SDPSCI_ED

Catalog No. S07747ND
Click to view available options
Quantity:
3/Container
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Anolis carolinensis is also know as the American chameleon.

Anolis is easy to keep and is interesting because of its habit of changing color.

  • With instructions

Specifications

Storage Requirements 24°C to 27°C
Quantity 3/Container
Size Small
Scientific Name Anolis carolinensis
Includes Instructions
Sufficient For Class of 30 students
Product Type Unsexed Small Anolis carolinensis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.