Learn More
Carolina™ Bergey's Manual of Determinative Bacteriology
SDPSCI_ED
Catalog No. s07431nd
Click to view available options
Quantity:
Ea.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Readers will find short general descriptions that encompass all organisms by groups, shape and size, Gram reaction, other pertinent morphological features, motility and flagella, relations to oxygen, basic type of metabolism, carbon and energy sources, habitat and ecology.
This reference volume also includes new genera and species, new combinations and new taxa published through the January 1992 issue of the Journal of Systematic Bacteriology. Includes discussions of difficulties in identification, keys or tables to genera and species, genus descriptions, synonyms, other nomenclatural changes and numerous illustrations.
  • Grades 9 to college
  • Soft cover
  • John G. Holt, PhD. 1994 (9th ed)
  • 816 pages
  • Based on the data contained in the four volume Bergey's Manual of Systematic Bacteriology
Specifications
Quantity Ea.
Includes Includes discussions of difficulties in identification, keys or tables to genera and species, genus descriptions, synonyms, other nomenclatural changes and numerous illustrations
Sufficient For Class of 30 students
Product Type Bergey's Manual of Determinative Bacteriology Book
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.