Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Coleus
SDPSCI_ED

Catalog No. s07550nd Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
Ea.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

An especially good specimen for starch-iodine reaction studies, to show absence of photosynthesis in non-green areas.

  • White variegated
  • Vigorous clone
  • Easy to propagate from stem cuttings
  • One Plant

Specifications

For Use With (Application) Good specimen for starch iodine reaction studies, to show absence of photosynthesis in non-green areas.
Storage Requirements 22°C
Quantity Ea.
Scientific Name Coleus blumei
Sufficient For Class of 30 students
Product Type Coleus blumei
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.