Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Daphnia magna
SDPSCI_ED

Cultures of D. magna contain adults, juveniles, and neonates.

$29.25

Specifications

For Use With (Application) Commonly used in aquatic effluent toxicity testing, obligate algal feeder.
Storage Requirements 21°C
Quantity Ea.
Scientific Name Daphnia magna
Includes Adults, juveniles, neonates
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S07268CND
SDPSCI_ED
View Documents
Carolina Biological Supply
TF142330LC
Daphnia Magna Coupon
Each for $29.25
Only null left
Add to Cart
 
S07268ND
SDPSCI_ED
View Documents
Carolina Biological Supply
142330
Daphnia Magna N/A N/A
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.
Description

Description

  • For a class of 30 students
  • A large species of Daphnia
  • Commonly used in aquatic effluent toxicity testing
  • An obligate algal feeder
  • Cultured using powdered Spirulina as a food source

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

Specifications

Commonly used in aquatic effluent toxicity testing, obligate algal feeder.
Ea.
Adults, juveniles, neonates
21°C
Daphnia magna
Class of 30 students
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.