Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Daphnia pulex
SDPSCI_ED

Catalog No. S07269ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S07269ND 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07269ND Supplier Carolina Biological Supply Supplier No. 142314

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Daphnia pulex is the most common species of water flea.

Daphnia pulex is an easy-to-grow food source for hydra and small fish.

  • For a class of 30 students
  • Recommended for environmental studies
  • Prolific: grow to feed your fish

Specifications

For Use With (Application) Daphnia pulex is an easy-to-grow food source for hydra and small fish.
Packaging Jar
Storage Requirements 21°C
Quantity 1 Ea.
Includes Several instars (including large forms with eggs), instructions
Sufficient For Class of 30 students
Product Type Daphnia Pulex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.