Learn More
Carolina™ Enterobacter cloacae, Living, Tube
SDPSCI_ED
Catalog No. s21897nd
Click to view available options
Quantity:
1 Tube
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
This culture is Gram-, motile rods, found in soil, water, and sewage.
It is cultured in nutrient agar at 30° C in a tube. Cultures are guaranteed as to species and purity.
  • Each culture contains enough material for a class of 30 students.
Specifications
Storage Requirements 30°C
Quantity 1 Tube
Media Type Nutrient Broth
Product Type Bacteria Culture
Gram Stain Result Gram(-)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.