Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Neisseria subflava, MicroKwik Culture™, Vial

Shop All Carolina Biological Supply Products


Click to view available options
:
Neisseria subflava, MicroKwik Culture
Description
Includes
Note: Each MicroKwik Culture™ comes with 5mL of rehydration medium.
Every culture is guaranteed for species and purity. Bacteria cultures are labeled by genus and species name, media on which they are cultivated, and temperature for optimal growth. All Gram staining is based on 24-hour cultures. Special packing and transportation costs are extra for animal and human pathogens.
- Each culture contains enough material for a class of 30 students
- It is cultured on brain heart infusion agar 37°C
- MicroKwik Culture™ is a Carolina™ trademark for freeze-dried, viable bacteria plus medium
- Since MicroKwik Cultures can be stored under refrigeration, they can be ordered ahead of time
Specifications
Specifications
Quantity | 1 Vial |
Media Type | Brain Heart Infusion Agar |
Product Type | Bacteria Culture |
Gram Stain Result | Gram (-) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction