Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Physarum Culture Kit
SDPSCI_ED

Catalog No. s07215nd Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
1 Ea.
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Includes

  • 30 Physarum Life Cycle Bioreview™ Sheets
  • Physarum polycephalum plate culture (plasmodial stage)
  • Physarum Sclerotium
  • 4 bottles agar, non-nutrient (2%)
  • 20 sterile Petri dishes
  • Old-fashoned oat flakes
  • Sterile disposable scalpel
  • Autoclave disposal bag

Despite its confusing taxonomic status, the creeping slime mold Physarum polycephalum is still an attention-getter for your students.

Students can follow the life cycle, observing the active plasmodial stage and the inactive sclerotial stage.

  • Easy to perform
  • Requires no experience in microbiology
  • For a class of 30

Note: Upon receipt of kit, break seal on plate culture and store at room temperature away from direct light. For best results, use plate culture within 2 to 3 days of receipt. Kit and perishable components will ship separately and may arrive on different days.

Specifications

For Use With (Application) Students can follow the life cycle, observing the active plasmodial stage and the inactive sclerotial stage.
Quantity 1 Ea.
Scientific Name Physarum
Includes 30 Physarum life cycle Bioreview™ sheets, Physarum polycephalum plate culture (plasmodial stage), Physarum Sclerotium, 4 bottles agar, non-nutrient (2%), 20 sterile petri dishes, old fashioned oat flakes, sterile disposable scalpel
Sufficient For Class of 30 students
Product Type Bacteria Culture Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.