Learn More
Carolina™ Individual Bacterial Cultures (A-C)



Description
Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.
- Labeled by genus and species name and media on which they are cultured
- All Gram staining based on 24 hr. cultures
- Pure and high-quality
- For every level of study
- Easy-to-use media
‡ This is a human or animal pathogen. It cannot be shipped to high schools, preparatory schools, or individuals, in accordance with guidelines of the U.S. Department of Health and Human Services.
Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.
Specifications
Specifications
For Use With (Application) | Ideal for food and chemical industries because of their highly diverse metabolic activities. |
Packaging | Tube |
Storage Requirements | 20°C to 22°C |
Quantity | Ea. |
Scientific Name | Bacillus stearothermophilus |
Includes | Carolina™ techniques for studying bacteria and fungi manual |
Sufficient For | Class of 30 students |
Media Type | Tryptic Soy Agar |
Product Type | Bacteria Culture |
Temperature (Metric) Range | 55°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.