Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Bacterial Cultures (D-M)
SDPSCI_ED

Catalog No. S07427ND Shop All Carolina Biological Supply Products
Click to view available options
Product Type:
Bacteria Culture
Bacteria Culture Plate
Bacteria Culture Tube
Bacteria MicroKwik Culture
Scientific Name:
E.coli
Enterobacter aerogenes
Enterococcus faecalis
Klebsiella pneumoniae
Lactococcus lactus
Micrococcus luteus
Micrococcus roseus
Mycobacterium phlei
Mycobacterium smegmatis
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.

Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.

  • Labeled by genus and species name and media on which they are cultured
  • All Gram staining based on 24 hr. cultures
  • Pure and high-quality
  • For every level of study
  • Easy-to-use media

‡ This is a human or animal pathogen. It cannot be shipped to high schools, preparatory schools, or individuals, in accordance with guidelines of the U.S. Department of Health and Human Services.

Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.

Specifications

For Use With (Application) Ideal for food and chemical industries because of their highly diverse metabolic activities.
Packaging Vial
Storage Requirements 20°C to 22°C
Quantity Ea.
Scientific Name Micrococcus luteus
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Media Type Brain Heart Infusion Agar/Broth
Product Type Bacteria MicroKwik Culture
Temperature (Metric) Range 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.